Learn More
Macherey-Nagel™ Nanaocolor™ Tube Test
Perform easy, fast measurements. Macherey-Nagel™ Nanocolor™ Tube Tests are bar coded for automatic recognition by Macherey-Nagel photometers. Each test includes exactly pre-dosed reagents in 16mm cuvettes and additional reagents for easy handling, high user safety and maximum precision.
Brand: Macherey-Nagel™ 985017
Additional Details : Weight : 0.50000kg
Description
Nanocolor™ tube tests for photometric analysis feature easy handling, making them the first choice for routine, laboratory and process analysis. They are ideally packed in stable boxes with color coded labels, giving all relevant information about the test at one glance. A variety of test kits are available with different measurement ranges for all typical parameters relevant in water and waste water analysis.
- Easy
- Colored pictograms as step-by-step instruction
- Big cuvettes for easy pipetting
- Barcoded cuvettes for automatic test selection
- Safe
- Convenient withdrawal of tubes from the box
- No contact with chemicals
- Reactions based on internationally accepted standard methods
- Reliable
- Precisely pre-dosed reagents
- Adequate test for every application
- Constant high quality from batch to batch
Specifications
Chlorine (Cl2), Ozone (O3) | |
Pink label | |
Photometric determination | |
20 test tubes Chlorine / Ozone 2, 1 bottle with 5mL Chlorine / Ozone 2 R2 | |
20 Pack | |
4.0 mL | |
1 min. (free), 3 min. (total) |
DIN, EN, ISO, EPA and APHA standards | |
Pre-dosed Cuvettes | |
Compatible Macherey-Nagel photometers | |
20 | |
0.05 - 2.50mg/L Cl2, 0.05 - 2.00mg/L O3 | |
DPD | |
Cuvette Test |