Learn More
Namodenoson, MedChemExpress
Product Code. 16491357
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
Product Code. 16491357
Brand: Medchem Express HY12365
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Namodenoson (CF-102) is a selective A3 adenosine receptor (A3AR) agonist (Ki=0.33 nM). Namodenoson displays 2500- and 1400-fold selectivity over A1 and A2A receptors respectively.
Namodenoson (CF-102) is a selective A3 adenosine receptor (A3AR) agonist (Ki=0.33 nM). Namodenoson displays 2500- and 1400-fold selectivity over A1 and A2A receptors respectively.
Specifications
Chemical Name or Material Namodenoson
CAS 163042-96-4
Color White
Purity Grade Notes Research
Molecular Formula C18H18ClIN6O4
Quantity 50 mg
Synonym CF-102 2-Cl-IB-MECA
Solubility Information DMSO : ≥ 31 mg/mL (56.91 mM)
SMILES O[C@H]([C@@H]1O)[C@@H](O[C@@H]1C(NC)=O)N2C3=NC(Cl)=NC(NCC4=CC(I)=CC=C4)=C3N=C2
Molecular Weight (g/mol) 544.73
Formula Weight 544.73
Percent Purity 99.6%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death