missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-S-trityl-D-cysteine, 98%, Thermo Scientific Chemicals

Catalog Number p-7041488
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
167015-11-4
C37H31NO4S
585.718
MFCD00151922
KLBPUVPNPAJWHZ-UUWRZZSWSA-N
fmoc-d-cys trt-oh, n-fmoc-s-trityl-d-cysteine, fmoc-s-trityl-d-cysteine, n-alpha-9-fluorenylmethyloxycarbonyl-s-trityl-d-cysteine, n-9h-fluoren-9-ylmethoxy carbonyl-s-triphenylmethyl-d-cysteine, d-cysteine, n-9h-fluoren-9-ylmethoxy carbonyl-s-triphenylmethyl, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-triphenylmethyl sulfanyl propanoic acid, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tritylthio propanoic acid, ambotzfaa1035, pubchem19012
7168037
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-tritylsulfanylpropanoic acid
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)SCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46
This item is not returnable. View return policy
167015-11-4
C37H31NO4S
585.718
MFCD00151922
KLBPUVPNPAJWHZ-UUWRZZSWSA-N
fmoc-d-cys trt-oh, n-fmoc-s-trityl-d-cysteine, fmoc-s-trityl-d-cysteine, n-alpha-9-fluorenylmethyloxycarbonyl-s-trityl-d-cysteine, n-9h-fluoren-9-ylmethoxy carbonyl-s-triphenylmethyl-d-cysteine, d-cysteine, n-9h-fluoren-9-ylmethoxy carbonyl-s-triphenylmethyl, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-triphenylmethyl sulfanyl propanoic acid, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tritylthio propanoic acid, ambotzfaa1035, pubchem19012
7168037
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-tritylsulfanylpropanoic acid
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)SCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 172°C to 176°C
Quantity 1 g
Beilstein 7558914
Formula Weight 587.71
Percent Purity 98%
Chemical Name or Material Nalpha-Fmoc-S-trityl-D-cysteine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'