missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-N^w-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-L-arginine, 98%

Product Code. p-7039685
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15475435 5 g 5g
15485435 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475435 Supplier Thermo Scientific Alfa Aesar Supplier No. H59039.06
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 154445-77-9 | C34H40N4O7S | 648.775 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154445-77-9
Molecular Formula C34H40N4O7S
Molecular Weight (g/mol) 648.775
MDL Number MFCD00235804
InChI Key HNICLNKVURBTKV-NDEPHWFRSA-N
Synonym fmoc-arg pbf-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-3-2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl sulfonyl guanidino pentanoic acid, nalpha-fmoc-nomega-pbf-l-arginine, fmoc-arg pbf, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, fmoc-arg pdf-oh, pubchem9946, fmoc-l-arg pbf-oh, fmoc-arg pbf-oh hplc, n-alpha-9-fluorenylmethoxycarbonyl-n-omega-2,2,4
PubChem CID 11354259
IUPAC Name (2S)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
SMILES CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)N)C)CC(O2)(C)C

Specifications

Quantity 5 g
Beilstein 8302671
Optical Rotation −5.5° (c=1 in DMF)
Formula Weight 648.77
Percent Purity 98%
Chemical Name or Material Nalpha-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-L-arginine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.