missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-Nepsilon-Fmoc-D-lysine, 95%

Product Code. p-7041441
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15408698 250 mg 250mg
15488688 1 g 1g
15498688 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 15408698

Brand: Thermo Scientific Alfa Aesar H62116.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 115186-31-7 | C26H32N2O6 | 468.55 g/mol

Nalpha-Boc-Nepsilon-Fmoc-D-lysine is commonly used in the synthesis of aza-amino acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha-Boc-Nepsilon-Fmoc-D-lysine is commonly used in the synthesis of aza-amino acids.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115186-31-7
Molecular Formula C26H32N2O6
Molecular Weight (g/mol) 468.55
MDL Number MFCD00076966
InChI Key JYEVQYFWINBXJU-ANBDAQEENA-N
Synonym boc-d-lys fmoc-oh, n-boc-n'-fmoc-d-lysine, boc-l-lys fmoc-oh, boc-n-epsilon-fmoc-d-lysine, n-alpha-t-butyloxycarbonyl-n-epsilon-9-fluorenylmethyloxycarbonyl-d-lysine, 2r-2-tert-butoxycarbonyl amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzbaa1040, n-boc-n-fmoc-d-lysine, pubchem12013, n2-boc-n6-fmoc-d-lysine
PubChem CID 14019424
IUPAC Name (2R)-6-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 117°C to 123°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 468.55
Percent Purity 95%
Chemical Name or Material Nalpha-Boc-Nepsilon-Fmoc-D-lysine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.