Learn More
Naloxone Hydrochloride, TRC
Product Code. 17911865
Change view
Click to view available options
Quantity:
100 mg
250 mg
5 g
Unit Size:
100mg
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17911865 5 g 5g
17912095 100 mg 100mg
17971445 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17911865 Supplier Toronto Research Chemicals Supplier No. TRCN2850005G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 357-08-4
Molecular Formula C19 H21 N O4 . Cl H
Molecular Weight (g/mol) 363.84
Synonym Morphinan-6-one, 4,5-epoxy-3,14-dihydroxy-17-(2-propen-1-yl)-, hydrochloride (1:1), (5α)-, Morphinan-6-one, 17-allyl-4,5α-epoxy-3,14-dihydroxy-, hydrochloride (8CI), Morphinan-6-one, 4,5-epoxy-3,14-dihydroxy-17-(2-propenyl)-, hydrochloride, (5α)- (9CI), Normorphinone, N-allyl-7,8-dihydro-14-hydroxy-, hydrochloride (7CI), (-)-N-Allyl-14-hydroxynordihydromorphinone hydrochloride, (-)-Naloxone hydrochloride, EN 1530, EN 15304, N-Allyldihydro-14-hydroxynormorphinone hydrochloride, NIH 7890, Nalone, Nalonee, Naloxone hydrochloride, Narcan, Narcanti, l-Naloxone hydrochloride
IUPAC Name (4R,4aS,7aR,12bS)-4a,9-dihydroxy-3-prop-2-enyl-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;hydrochloride
SMILES Cl.Oc1ccc2C[C@H]3N(CC=C)CC[C@@]45[C@@H](Oc1c24)C(=O)CC[C@@]35O
Specifications
Quantity 5 g
InChI Formula InChI=1S/C19H21NO4.ClH/c1-2-8-20-9-7-18-15-11-3-4-12(21)16(15)24-17(18)13(22)5-6-19(18,23)14(20)10-11;/h2-4,14,17,21,23H,1,5-10H2;1H/t14-,17+,18+,19-;/m1./s1
Formula Weight 363.1237
Percent Purity >95%
Chemical Name or Material Naloxone Hydrochloride
Grade HPLC