Learn More
Thermo Scientific™ Nalgene™ Staining Box
Description
Stain, fix and handle fragile electrophoresis gels and membranes with less chance of damage than conventional methods. Thermo Scientific™ Nalgene™ Staining Box is equipped with a bottom drain allowing you to change solution without disturbing the box's other fragile contents. Box holds 4.8 in. x 4.8 in (189 mm x 189 mm) gel along with 500mL of liquid.
- Polymethylpentene box and polypropylene copolymer cover
- Useful over a -70° to +70°C (-94° to +158°F) temperature range
- Good chemical compatibility with acids, bases and select organic solvents for a versatile range of protocols
- LDPE plug in bottom for easy drainage of liquid with minimal handling of gel and provides a leakproof seal
- Plug won't interfere with cover or contents of boxes on a benchtop or shaker
- Stackable, with a handle on each end for safe, easy carrying
- Microwavable and easy to clean
Specifications
Specifications
| Description | Replacement Plug for 5705-1010 and -2020 |
| Type | Staining Box Plug |
| No. per Case | 12 |
| No. per Pack | 12 |