missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Naked1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517389
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
In the mouse, Nkd is a Dishevelled (see DVL1; MIM 601365)-binding protein that functions as a negative regulator of the Wnt (see WNT1; MIM 164820)-beta-catenin-Tcf signaling pathway.
Specifications
| Naked1 | |
| Polyclonal | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| Q969G9 | |
| NKD1 | |
| Synthetic peptide corresponding to residues surrounding Gln227 of human Naked1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| NKD1 | |
| 2810434J10Rik; 9030215G15Rik; Dvl-binding protein; hNkd; hNkd1; mNkd; mNkd1; naked cuticle 1 homolog; naked cuticle 1 homolog (Drosophila); naked cuticle homolog 1; naked cuticle homolog 1 (Drosophila); naked cuticle-1; Naked1; naked-1; Nkd; NKD1; PP7246; Protein naked cuticle homolog 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 85407 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction