Learn More
NAGA Monoclonal Antibody (OTI6F3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811239S
Description
NAGA Monoclonal antibody specifically detects NAGA in Human samples. It is validated for Western BlotSpecifications
| NAGA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q03181 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5467 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI6F3 | |
| Unconjugated | |
| Ppard | |
| Acetylgalactosaminidase, alpha-N- (alpha-galactosidase B); alpha-galactosidase B; Alpha-N-acetylgalactosaminidase; D22S674; EC 3.2.1; EC 3.2.1.49; galactosidasealphaB; galactosidase-alphaB; GALB; N-acetyl galactosaminidase, alpha; N-acetylgalactosaminidase, alpha-; Naga | |
| Human recombinant protein fragment corresponding to amino acids 1-244 of human PPARD produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.