Learn More
Nafoxidine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Nafoxidine Hydrochloride |
| CAS | 1847-63-8 |
| Molecular Formula | C29 H31 N O2 . Cl H |
| Quantity | 5 mg |
| Synonym | 1-[2-[4-(3,4-dihydro-6-methoxy-2-phenyl-1-naphthalenyl)phenoxy]ethyl]-, hydrochloride (1:1)Pyrrolidine, 1-[2-[4-(3,4-dihydro-6-methoxy-2-phenyl-1-naphthalenyl)phenoxy]ethyl]-, hydrochloride (9CI), Pyrrolidine, 1-[2-[p-(3,4-dihydro-6-methoxy-2-phenyl-1-naphthyl)phenoxy]ethyl]-, hydrochloride (7CI,8CI), 1-[2-[p-(3,4-Dihydro-6-methoxy-2-phenyl-1-naphthyl)phenoxy]ethyl]pyrrolidine hydrochloride, 11100A, NSC 70735, Nafoxidine hydrochloride, U 11100, U 11100A |
| InChI Formula | InChI=1S/C29H31NO2.ClH/c1-31-26-14-16-28-24(21-26)11-15-27(22-7-3-2-4-8-22)29(28)23-9-12-25(13-10-23)32-20-19-30-17-5-6-18-30;/h2-4,7-10,12-14,16,21H,5-6,11,15,17-20H2,1H3;1H |
| SMILES | Cl.COc1ccc2C(=C(CCc2c1)c3ccccc3)c4ccc(OCCN5CCCC5)cc4 |
| IUPAC Name | 1-[2-[4-(6-methoxy-2-phenyl-3,4-dihydronaphthalen-1-yl)phenoxy]ethyl]pyrrolidine;hydrochloride |
| Molecular Weight (g/mol) | 462.02 |
| Formula Weight | 461.2122 |