missing translation for 'onlineSavingsMsg'
Learn More

NADPH tetracyclohexanamine, MedChemExpress

Product Code. p-200101804
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16468778

Brand: Medchem Express HYF0003A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NADPH tetracyclohexanamine is a ubiquitous cofactor and biological reducing agent.

NADPH tetracyclohexanamine is a ubiquitous cofactor and biological reducing agent.

Specifications

Chemical Name or Material NADPH tetracyclohexanamine
CAS 100929-71-3
Color White
Purity Grade Notes Research
Molecular Formula C45H82N11O17P3
Quantity 10 mg
Solubility Information H2O : ≥ 100 mg/mL (87.56 mM) ∣DMSO : ≥ 84 mg/mL (73.55 mM)
SMILES NC1=C2C(N([C@H]3[C@H](OP(O)(O)=O)[C@H](O)[C@@H](COP(OP(O)(OC[C@@H]4[C@@H](O)[C@@H](O)[C@H](N5C=C(C(N)=O)CC=C5)O4)=O)(O)=O)O3)C=N2)=NC=N1.NC6CCCCC6.NC7CCCCC7.NC8CCCCC8.NC9CCCCC9
Molecular Weight (g/mol) 1142.12
Formula Weight 1142.12
Percent Purity 96.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.