missing translation for 'onlineSavingsMsg'
Learn More

NABP1 Monoclonal Antibody (OTI4D5), TrueMAB™, OriGene

Product Code. 17564714
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17564714 30 μL 30µL
17544714 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17564714 Supplier Origene Technologies Supplier No. TA811086S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NABP1 Monoclonal antibody specifically detects NABP1 in Human samples. It is validated for Western Blot

Specifications

Antigen NABP1
Applications Western Blot
Classification Monoclonal
Clone OTI4D5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SPATA46
Gene Accession No. Q5T0L3
Gene Alias 4930434H03Rik; 4930442A21Rik; 4930488J04Rik; 4933440J18Rik; 5830411E10Rik; AI852561; hSSB2; Nabp1; Nbp1; nucleic acid binding protein 1; nucleic acid-binding protein 1; Obfc2a; oligonucleotide/oligosaccharide-binding fold containing 2A; oligonucleotide/oligosaccharide-binding fold-containing protein 2A; RGD1306658; Sensor of single-strand DNA complex subunit B2; sensor of ssDNA subunit B2; Single-stranded DNA-binding protein 2; SOSS complex subunit B2; SOSS-B2; Ssb2
Host Species Mouse
Immunogen Full length human recombinant protein of human C1ORF111 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284680
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.