Learn More
N6-methyladenosine (m6A) Antibody (7N6C7) [HRP], Novus Biologicals Biologicals™
Product Code. 30499838 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30499838 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30499838 Supplier Novus Biologicals Supplier No. NBP333194H
Please to purchase this item. Need a web account? Register with us today!
Mouse Monoclonal Antibody
N6-methyladenosine (m6A) Monoclonal antibody specifically detects N6-methyladenosine (m6A) in Non-species specific samples. It is validated for ELISA,Dot Blot
TRUSTED_SUSTAINABILITY
Specifications
Antigen N6-methyladenosine (m6A)
Applications ELISA, Dot Blot
Classification Monoclonal
Clone 7N6C7
Conjugate HRP
Formulation PBS
Gene Alias (-)-6-(Methylamino)purine riboside, N(6)-Methyladenosine, N-Methyladenosine
Host Species Mouse
Immunogen Chemical compounds corresponding to N6-methyladenosine (m6A).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ