Learn More
N6-[(1,1-Dimethylethoxy)carbonyl]-N2-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine, TRC
Specifications
Specifications
| CAS | 92122-45-7 |
| Molecular Formula | C26H32N2O6 |
| Quantity | 1 g |
| Synonym | (R)-6-[(tert-Butoxycarbonyl)amino]-2-[[[(9H-fluoren-9-yl)methoxy]carbonyl]amino]hexanoic Acid, Fmoc-D-Lys(Boc)-OH, Nalpha-Fmoc-Nepsilon-Boc-D-lysine |
| InChI Formula | InChI=1S/C26H32N2O6/c1-26(2,3)34-24(31)27-15-9-8-14-22(23(29)30)28-25(32)33-16-21-19-12-6-4-10-17(19)18-11-5-7-13-20(18)21/h4-7,10-13,21-22H,8-9,14-16H2,1-3H3,(H,27,31)(H,28,32)(H,29,30)/t22-/m1/s1 |
| SMILES | CC(C)(C)OC(=O)NCCCC[C@@H](NC(=O)OCC1c2ccccc2c3ccccc13)C(=O)O |
| Molecular Weight (g/mol) | 468.54 |
| Formula Weight | 468.226 |
| Chemical Name or Material | N6-[(1,1-Dimethylethoxy)carbonyl]-N2-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine |