missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
N4BP2L1 Polyclonal antibody specifically detects N4BP2L1 in Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | N4BP2L1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | CG018, CG081, NEDD4 binding protein 2-like 1, NEDD4-binding protein 2-like 1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 40-120 of human N4BP2L1 (NP_001073159). FRKHLYLLRGLPGSGKTTLARQLQHDFPRALIFSTDDFFFREDGAYEFNPDFLEEAHEWNQKRARKAMRNGISPIIIDNTN |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?