Learn More
N2-Methyl-2’-deoxyguanosine, TRC
Product Code. 30480161
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480161 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30480161 Supplier Toronto Research Chemicals Supplier No. TRCM29303525MG
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N2-Methyl-2'-deoxyguanosine
CAS 19916-77-9
Molecular Formula C11H15N5O4
Quantity 25 mg
Synonym 2'-Deoxy-N-methylguanosine, INX 09146, N-Methyl-2'-deoxyguanosine
InChI Formula InChI=1S/C11H15N5O4/c1-12-11-14-9-8(10(19)15-11)13-4-16(9)7-2-5(18)6(3-17)20-7/h4-7,17-18H,2-3H2,1H3,(H2,12,14,15,19)/t5-,6+,7+/m0/s1
SMILES OC[C@H]1O[C@@H](N(C=N2)C3=C2C(NC(NC)=N3)=O)C[C@@H]1O
IUPAC Name 9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-(methylamino)-1H-purin-6-one
Molecular Weight (g/mol) 281.27
Formula Weight 281.1124