Learn More
Tocris Bioscience™ N1,N11-Diethylnorspermine tetrahydrochloride
GREENER_CHOICE
Product Code. 18005462
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18005462
Brand: Tocris Bioscience™ 0468/10
Please to purchase this item. Need a web account? Register with us today!
Spermine and spermidine acetyltransferase potentiator
Highly potent inducer of spermidine/spermine N1-acetyltransferase, able to increase enzyme activity 200-1000 fold.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 156886-85-0
Molecular Formula C13H36Cl4N4
Specifications
Quantity 10 mg
Formula Weight 390.27
Purity >98%
Molecular Weight (g/mol) 390.259
Chemical Name or Material N1, N11-Diethylnorspermine tetrahydrochloride