missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Undecanoyl Chloride, TRC
Click to view available options
Quantity:
500 mg
10 g
Unit Size:
10g
500mg
Specifications
Specifications
| Chemical Name or Material | n-Undecanoyl Chloride |
| CAS | 17746-05-3 |
| Molecular Formula | C11 H21 Cl O |
| Quantity | 10 g |
| Synonym | NSC 83585, Undecanoic Acid Chloride |
| InChI Formula | InChI=1S/C11H21ClO/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2-10H2,1H3 |
| SMILES | CCCCCCCCCCC(=O)Cl |
| IUPAC Name | undecanoyl chloride |
| Molecular Weight (g/mol) | 204.737 |
| Formula Weight | 204.128 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction