Learn More
N-Trityl Losartan-d4, TRC
Specifications
Specifications
| Chemical Name or Material | N-Trityl Losartan-d4 |
| CAS | 1309283-24-6 |
| Molecular Formula | C41H33D4ClN6O |
| Quantity | 10 mg |
| Synonym | 2-Butyl-4-chloro-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl-d4]-4-yl]methyl]-1H-imidazole-5-methanol, Trityllosartan-d4 |
| InChI Formula | InChI=1S/C41H37ClN6O/c1-2-3-23-38-43-39(42)37(29-49)47(38)28-30-24-26-31(27-25-30)35-21-13-14-22-36(35)40-44-46-48(45-40)41(32-15-7-4-8-16-32,33-17-9-5-10-18-33)34-19-11-6-12-20-34/h4-22,24-27,49H,2-3,23,28-29H2,1H3/i24D,25D,26D,27D |
| SMILES | [2H]c1c([2H])c(c([2H])c([2H])c1Cn2c(CCCC)nc(Cl)c2CO)c3ccccc3c4nnn(n4)C(c5ccccc5)(c6ccccc6)c7ccccc7 |
| IUPAC Name | [2-butyl-5-chloro-3-[[2,3,5,6-tetradeuterio-4-[2-(2-trityltetrazol-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methanol |
| Molecular Weight (g/mol) | 669.25 |
| Formula Weight | 668.2968 |