Learn More
n-Tetradecyl-d29 Alcohol, CDN
Specifications
Specifications
| Chemical Name or Material | 1-Tetradecanol-D29 |
| CAS | 284474-78-8 |
| Molecular Formula | C14 D29 H O |
| Quantity | 0.25 g |
| Synonym | 1-Tetradecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-d29-ol (9 CI, ACI), 1-Tetradecanol-d29, Myristyl alcohol-d29, n-Tetradecyl-d29 Alcohol |
| InChI Formula | InChI=1 S/C14H30O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h15H,2-14H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])O |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-nonacosadeuteriotetradecan-1-ol |
| Molecular Weight (g/mol) | 243.5661 |
| Formula Weight | 243.4117 g/mol |
| Show More |