Learn More
N-tert-Butylisopropylamine, 98%
Product Code. 11443447
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443447 25 g 25g
11433447 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11443447 Supplier Thermo Scientific Alfa Aesar Supplier No. A17221.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 7515-80-2 | C7H18N | 116.23 g/mol
N-tert-Butylisopropylamine, is used as a participates in the synthesis of hindered enamines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-tert-Butylisopropylamine, is used as a participates in the synthesis of hindered enamines.
Solubility
It has optimal solubility in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7515-80-2
Molecular Formula C7H18N
Molecular Weight (g/mol) 116.23
MDL Number MFCD00037059
InChI Key ZWXQPERWRDHCMZ-UHFFFAOYSA-O
Synonym n-tert-butylisopropylamine, 2-propanamine, 2-methyl-n-1-methylethyl, tert-butylisopropylamine, n-isopropyl-2-methylpropan-2-amine, ethanamine, 1,1-dimethyl, n-1-methylethyl, n-isopropyl-tert-butylamine, tert-butyl methylethyl amine, tert-butyl isopropyl amine
PubChem CID 82023
IUPAC Name 2-methyl-N-propan-2-ylpropan-2-amine
SMILES CC(C)[NH2+]C(C)(C)C
Specifications
Density 0.727
Boiling Point 98°C
Flash Point −3°C (26°F)
Refractive Index 1.398
Quantity 25 g
UN Number UN2733
Beilstein 1846752
Solubility Information It has optimal solubility in water.
Formula Weight 115.22
Percent Purity 98%
Chemical Name or Material N-tert-Butylisopropylamine