Learn More
N-(Perfluoro-1-octanesulfonyl) Glycine (>80%), TRC
Specifications
Specifications
| Chemical Name or Material | N-(Perfluoro-1-octanesulfonyl) Glycine |
| CAS | 2806-24-8 |
| Molecular Formula | C10 H4 F17 N O4 S |
| Quantity | 25 mg |
| Synonym | Perfluorooctanesulfonamidoacetic Acid, Glycine','N-[(heptadecafluorooctyl)sulfonyl]- (6CI,8CI,9CI), N-[(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluorooctyl)sulfonyl]glycine, 2-(Perfluorooctanesulfonamido)acetic acid, Perfluorooctanesulfonamidoacetic acid, Glycine','N-[(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctyl)sulfonyl]-, FOSAA, N-[(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluorooctyl)sulfonyl]-glycine, N-[(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluorooctyl)sulfonyl]glycine, FOSAA |
| InChI Formula | InChI=1S/C10H4F17NO4S/c11-3(12,5(15,16)7(19,20)9(23,24)25)4(13,14)6(17,18)8(21,22)10(26,27)33(31,32)28-1-2(29)30/h28H,1H2,(H,29,30) |
| SMILES | OC(=O)CNS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC Name | 2-(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctylsulfonylamino)acetic acid |
| Molecular Weight (g/mol) | 557.18 |
| Formula Weight | 556.959 |