missing translation for 'onlineSavingsMsg'
Learn More

N-Palmitoyl-O-phosphocholine Serine-D9, TRC

Product Code. 30423898
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423898 10 mg 10mg
30428374 1 mg 1mg
30412471 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30423898 Supplier Toronto Research Chemicals Supplier No. TRCP34792110MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Palmitoyl-O-phosphocholine Serine-D9
Molecular Formula C24 D9 H40 N2 O7 P
Quantity 10 mg
Synonym (S)-2-Carboxy-2-palmitamidoethyl (2-(Tris(methyl-d3)ammonio)ethyl)phosphate
InChI Formula InChI=1S/C24H49N2O7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-23(27)25-22(24(28)29)21-33-34(30,31)32-20-19-26(2,3)4/h22H,5-21H2,1-4H3,(H2-,25,27,28,29,30,31)/t22-/m0/s1/i2D3,3D3,4D3
SMILES [2H]C([2H])([2H])[N+](CCOP(=O)([O-])OC[C@H](NC(=O)CCCCCCCCCCCCCCC)C(=O)O)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC Name [(2S)-2-carboxy-2-(hexadecanoylamino)ethyl] 2-[tris(trideuteriomethyl)azaniumyl]ethyl phosphate
Molecular Weight (g/mol) 517.684
Formula Weight 517.384
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.