missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N(omega)-Boc-N(beta)-Fmoc-L-beta-homolysine, 95%

Catalog Number p-7034503
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
203854-47-1
C27H34N2O6
482.58
MFCD01863054
SDBUQLGECIYUDT-LGWFVXIRNA-N
fmoc-l-beta-homolysine boc, s-3-9h-fluoren-9-yl methoxy carbonyl amino-7-tert-butoxycarbonyl amino heptanoic acid, fmoc-beta-holys boc-oh, fmoc-beta-homolys boc-oh, fmoc-l-nw-boc-beta-homolysine, 3s-7-1,1-dimethylethoxy carbonyl amino-3-9h-fluoren-9-ylmethoxy carbonyl amino heptanoic acid, 3s-7-tert-butoxycarbonyl amino-3-9h-fluoren-9-ylmethoxy carbonyl amino heptanoic acid, fmoc-l-beta-homolysine, ambotzfaa6700, fmoc-b-holys boc-oh
2761531
(3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-7-[(2-methylpropan-2-yl)oxycarbonylamino]heptanoic acid
CC(C)(C)OC(=O)NCCCC[C@@H](CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
This item is not returnable. View return policy
203854-47-1
C27H34N2O6
482.58
MFCD01863054
SDBUQLGECIYUDT-LGWFVXIRNA-N
fmoc-l-beta-homolysine boc, s-3-9h-fluoren-9-yl methoxy carbonyl amino-7-tert-butoxycarbonyl amino heptanoic acid, fmoc-beta-holys boc-oh, fmoc-beta-homolys boc-oh, fmoc-l-nw-boc-beta-homolysine, 3s-7-1,1-dimethylethoxy carbonyl amino-3-9h-fluoren-9-ylmethoxy carbonyl amino heptanoic acid, 3s-7-tert-butoxycarbonyl amino-3-9h-fluoren-9-ylmethoxy carbonyl amino heptanoic acid, fmoc-l-beta-homolysine, ambotzfaa6700, fmoc-b-holys boc-oh
2761531
(3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-7-[(2-methylpropan-2-yl)oxycarbonylamino]heptanoic acid
CC(C)(C)OC(=O)NCCCC[C@@H](CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

N(μ)-Boc-N(beta)-Fmoc-L-beta-homolysine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N(µ)-Boc-N(beta)-Fmoc-L-beta-homolysine is used as pharmaceutical intermediate.

Solubility
Sparingly soluble bin water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 8022293
Solubility Information Sparingly soluble bin water.
Formula Weight 482.58
Percent Purity 95%
Chemical Name or Material N(omega)-Boc-N(beta)-Fmoc-L-beta-homolysine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'