Learn More
MedChemExpress N-Nonyldeoxynojirimycin
Specifications
Specifications
| Purity | 99.0% |
| CAS | 81117-35-3 |
| Content And Storage | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Molecular Weight (g/mol) | 289.41 g/mol |
| Product Type | Peptides |
| Quantity | 5 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : 50 mg/mL (172.77 mM∣Need ultrasonic) |
| Synonym | NN-DNJ∣ Nonyl-DNJ |