Learn More
N-Nitrosoethylmethylamine-d3 (Major), TRC
Specifications
Specifications
| Chemical Name or Material | N-Nitrosoethylmethylamine-d3 (Major) |
| CAS | 69278-54-2 |
| Molecular Formula | C3D3H5N2O |
| Quantity | 5 mg |
| Synonym | N-Nitroso-methylethylamine D3 (Methyl D3), N-Methyl-N-nitrosoethanamine-d3, N-Nethyl-N-nitrosoethylamine-d3, Ethylmethylnitrosamine-d3, Methylethylnitrosamine-d3, Methylethylnitrosoamine-d3, N-Methyl-N-nitrosoethylamine-d3, N-Nitroso-N-methylethylamine-d3, N-Nitrosomethylethylamine-d3, NEMA-d3, Nitrosomethylethylamine-d3 |
| InChI Formula | InChI=1S/C3H8N2O/c1-3-5(2)4-6/h3H2,1-2H3/i2D3 |
| SMILES | [2H]C([2H])([2H])N(CC)N=O |
| IUPAC Name | N-ethyl-N-(trideuteriomethyl)nitrous amide |
| Molecular Weight (g/mol) | 91.13 |
| Formula Weight | 91.082 |