Learn More
N-Nitroso Varenicline, TRC
Product Code. 30136834
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136834 250 mg 250mg
30003569 25 mg 25mg
30004265 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136834 Supplier Toronto Research Chemicals Supplier No. TRCN249000250MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2755871-02-2
Molecular Formula C13H7D5N4O
Molecular Weight (g/mol) 245.3
Synonym 6,10-Methano-6H-pyrazino[2,3-h][3]benzazepine, 7,8,9,10-tetrahydro-8-nitroso- (ACI), 7,8,9,10-Tetrahydro-8-nitroso-6,10-methano-6H-pyrazino[2,3-h][3]benzazepine (ACI), N-Nitroso-Varenicline, N-Nitrosovarenicline, N-Nitroso Varenicline-d5
IUPAC Name 8-Nitroso-7,8,9,10-tetrahydro-6H-6,10-methanoazepino[4,5-g]quinoxaline-2,7,7,9,9-d5
SMILES O=NN1C[C@H]2C[C@@H](C1)c3cc4nccnc4cc23
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C13H12N4O/c18-16-17-6-8-3-9(7-17)11-5-13-12(4-10(8)11)14-1-2-15-13/h1-2,4-5,8-9H,3,6-7H2/t8-,9+
Formula Weight 245.13
Percent Purity >95%
Chemical Name or Material N-Nitrosovarenicline
Grade HPLC