missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitroso Tofenacin-d5, TRC
Specifications
Specifications
| Chemical Name or Material | N-Nitroso Tofenacin-d5 |
| Molecular Formula | C17 D5 H15 N2 O2 |
| Quantity | 5 mg |
| Synonym | N-Methyl-N-(2-(phenyl(o-tolyl)methoxy)ethyl)nitrous Amide-d5, N-Nitroso N-Demethylorphenadrine-d5, N-Nitroso N-Desmethylorphenadrine-d5, N-Nitroso N-Methyl-2-[(o-methyl-α-phenylbenzyl)oxy]ethylamine-d5 |
| InChI Formula | InChI=1S/C17H20N2O2/c1-14-8-6-7-11-16(14)17(15-9-4-3-5-10-15)21-13-12-19(2)18-20/h3-11,17H,12-13H2,1-2H3/i3D,4D,5D,9D,10D |
| SMILES | [2H]c1c([2H])c([2H])c(C(OCCN(C)N=O)c2ccccc2C)c([2H])c1[2H] |
| IUPAC Name | N-methyl-N-[2-[(2-methylphenyl)-(2,3,4,5,6-pentadeuteriophenyl)methoxy]ethyl]nitrous amide |
| Molecular Weight (g/mol) | 289.384 |
| Formula Weight | 289.184 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?