Learn More
N-Nitroso-N-methylaniline, TRC
Product Code. 18094030
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18094030 100 mg 100mg
18081550 50 mg 50mg
18082950 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18094030 Supplier Toronto Research Chemicals Supplier No. TRCN529925100MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 614-00-6
Molecular Formula C7 H8 N2 O
Molecular Weight (g/mol) 136.15
Synonym Aniline, N-methyl-N-nitroso- (6CI,8CI), N-Methyl-N-nitrosobenzenamine, Methylphenylnitrosamine, Methylphenylnitrosoamine, N-Methyl-N-nitrosoaniline, N-Methyl-N-phenylnitrosamine, N-Methyl-N-phenylnitrosoamine, N-Nitroso-N-methylaniline, N-Nitroso-N-methylphenylamine, NSC 137, Nitrosomethylphenylamine, Phenylmethylnitrosamine
IUPAC Name N-methyl-N-phenylnitrous amide
SMILES CN(N=O)c1ccccc1
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C7H8N2O/c1-9(8-10)7-5-3-2-4-6-7/h2-6H,1H3
Formula Weight 136.0637
Percent Purity >95%
Chemical Name or Material N-Nitroso-N-methylaniline
Grade HPLC