Learn More
N-Nitroso Labetalol, TRC
Specifications
Specifications
| Chemical Name or Material | N-Nitroso Labetalol |
| CAS | 2820170-74-7 |
| Molecular Formula | C19 H23 N3 O4 |
| Quantity | 50 mg |
| Synonym | 2-Hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenyl-propyl)-nitroso-amino]ethyl]benzamide, N-Nitroso Labetalol, 2-Hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)nitrosoamino]ethyl]benzamide, Benzamide','2-hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)nitrosoamino]ethyl]- |
| InChI Formula | InChI=1S/C19H23N3O4/c1-13(7-8-14-5-3-2-4-6-14)22(21-26)12-18(24)15-9-10-17(23)16(11-15)19(20)25/h2-6,9-11,13,18,23-24H,7-8,12H2,1H3,(H2,20,25) |
| SMILES | O=NN(C(CCC1=CC=CC=C1)C)CC(C2=CC=C(C(C(N)=O)=C2)O)O |
| IUPAC Name | 2-hydroxy-5-[1-hydroxy-2-[nitroso(4-phenylbutan-2-yl)amino]ethyl]benzamide |
| Molecular Weight (g/mol) | 357.4036 |
| Formula Weight | 357.1689 |