Learn More
N-Nitroso-bisoprolol, TRC
Specifications
Specifications
| Chemical Name or Material | N-Nitroso-bisoprolol |
| CAS | 2820170-76-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18H30N2O5 |
| Quantity | 100 mg |
| Synonym | N-Nitroso Bisoprolol |
| InChI Formula | InChI=1S/C18H30N2O5/c1-14(2)20(19-22)11-17(21)13-25-18-7-5-16(6-8-18)12-23-9-10-24-15(3)4/h5-8,14-15,17,21H,9-13H2,1-4H3 |
| SMILES | O=NN(CC(O)COC1=CC=C(C=C1)COCCOC(C)C)C(C)C |
| IUPAC Name | N-(2-hydroxy-3-(4-((2-isopropoxyethoxy)methyl)phenoxy)propyl)-N-isopropylnitrous amide |
| Molecular Weight (g/mol) | 354.44 |
| Show More |