Learn More
N,N,N'-Trimethyl-1,4-benzenediamine Dihydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | N,N,N'-Trimethyl-1,4-benzenediamine Dihydrochloride |
| CAS | 2739-08-4 |
| Molecular Formula | C9H14N2 .2HCl |
| Quantity | 100 mg |
| Synonym | N1,N1,N4-Trimethylbenzene-1,4-diamine, 4-(Dimethylamino)-N-methylaniline Dihydrochloride, N,N,N'-Trimethyl-1,4-phenylenediamine Dihydrochloride, N,N,N'-Trimethyl-p-phenylenediamine Dihydrochloride |
| InChI Formula | InChI=1S/C9H14N2/c1-10-8-4-6-9(7-5-8)11(2)3/h4-7,10H,1-3H3 |
| SMILES | CNc1ccc(cc1)N(C)C |
| IUPAC Name | 1-N,1-N,4-N-trimethylbenzene-1,4-diamine |
| Molecular Weight (g/mol) | 150.22 |
| Formula Weight | 150.1157 |