Learn More
N,N-Dimethylformamide-(carbonyl-13C), TRC
Specifications
Specifications
| Chemical Name or Material | N,N-Dimethylformamide carbonyl-13C |
| CAS | 32488-43-0 |
| Molecular Formula | 13C C2 H7 N O |
| Quantity | 25 mg |
| Synonym | N,N-Dimethylformamide (carbonyl-13C) |
| InChI Formula | InChI=1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3/i3+1 |
| SMILES | CN(C)[13CH]=O |
| IUPAC Name | N,N-dimethylformamide |
| Molecular Weight (g/mol) | 74.09 |
| Formula Weight | 74.0561 |