missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylbutyramide, 98%

Product Code. 11317495
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317495 5 g 5g
11327495 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317495 Supplier Thermo Scientific Alfa Aesar Supplier No. L05556.06
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 760-79-2 | C6H13NO | 115.176 g/mol

N,N-Dimethylbutyramide, is used as an organic intermediate, pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylbutyramide, is used as an organic intermediate, pharmaceutical intermediate.

Solubility
Very Soluble in water (110 g/L) (25°C).

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 760-79-2
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00015222
InChI Key VIJUZNJJLALGNJ-UHFFFAOYSA-N
Synonym n,n-dimethylbutyramide, butanamide, n,n-dimethyl, n,n-dimethyl-n-butyramide, nn-dimethylbutyramide, butyramide, n,n-dimethyl, butanamide,n-dimethyl, butanamide,n,n-dimethyl, butanoic acid dimethylamide, nn-dimethylbutyramide, pract., wln: 3vn1&1
PubChem CID 12973
IUPAC Name N,N-dimethylbutanamide
SMILES CCCC(=O)N(C)C

Specifications

Boiling Point 80°C to 82°C (15 mmHg)
Refractive Index 1.4425
Quantity 5 g
Beilstein 1742172
Solubility Information Very Soluble in water (110g/L) (25°C).
Formula Weight 115.17
Percent Purity 98%
Chemical Name or Material N,N-Dimethylbutyramide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.