missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-d6-glycine Hydrochloride, TRC

Product Code. 30370396
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30370396 10 mg -
30388708 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30370396 Supplier Toronto Research Chemicals Supplier No. TRCD47308210MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 347840-03-3
Molecular Formula C4 2H6 H3 N O2 . Cl H
Molecular Weight (g/mol) 145.62
Synonym Glycine, N,N-di(methyl-d3)-, hydrochloride (9CI), (Dimethylamino)acetic acid-d6 hydrochloride, 2-[Di(methyl-d3)amino]acetic acid hydrochloride, Dimethylglycine-d6 hydrochloride, N,N-Di(methyl-d3)aminoacetic acid hydrochloric acid, N,N-Dimethylglycine-d6 hydrochloride, N,N-Dimethylglycine-D6 (di(methyl-D3)) hydrochloride
IUPAC Name 2-[bis(trideuteriomethyl)amino]acetic acid;hydrochloride
SMILES Cl.[2H]C([2H])([2H])N(CC(=O)O)C([2H])([2H])[2H]

Specifications

InChI Formula InChI=1S/C4H9NO2.ClH/c1-5(2)3-4(6)7;/h3H2,1-2H3,(H,6,7);1H/i1D3,2D3;
Quantity 10 mg
Formula Weight 145.0777
Shipping Condition Room Temperature
Chemical Name or Material N,N-Dimethyl-d6-glycine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.