Learn More
N,N-Dimethyl-d6-glycine Hydrochloride, TRC
Product Code. 30388708
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388708 100 mg 100mg
30370396 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30388708 Supplier Toronto Research Chemicals Supplier No. TRCD473082100MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 347840-03-3
Molecular Formula C4 2H6 H3 N O2 . Cl H
Molecular Weight (g/mol) 145.62
Synonym Glycine, N,N-di(methyl-d3)-, hydrochloride (9CI), (Dimethylamino)acetic acid-d6 hydrochloride, 2-[Di(methyl-d3)amino]acetic acid hydrochloride, Dimethylglycine-d6 hydrochloride, N,N-Di(methyl-d3)aminoacetic acid hydrochloric acid, N,N-Dimethylglycine-d6 hydrochloride, N,N-Dimethylglycine-D6 (di(methyl-D3)) hydrochloride
IUPAC Name 2-[bis(trideuteriomethyl)amino]acetic acid;hydrochloride
SMILES Cl.[2H]C([2H])([2H])N(CC(=O)O)C([2H])([2H])[2H]
Specifications
InChI Formula InChI=1S/C4H9NO2.ClH/c1-5(2)3-4(6)7;/h3H2,1-2H3,(H,6,7);1H/i1D3,2D3;
Quantity 100 mg
Formula Weight 145.0777
Shipping Condition Room Temperature
Chemical Name or Material N,N-Dimethyl-d6-glycine hydrochloride