missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-2-(1-piperazinyl)nicotinamide hydrochloride, 96%, Thermo Scientific™

Product Code. 15409255
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15409255 1 g 1g
15419255 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15409255 Supplier Thermo Scientific Alfa Aesar Supplier No. H51679.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 902836-08-2
Molecular Formula C12H19ClN4O
Molecular Weight (g/mol) 270.761
MDL Number MFCD12407084
InChI Key YRPLJKUUYNCGQS-UHFFFAOYSA-N
Synonym n,n-dimethyl-2-piperazin-1-yl nicotinamide hydrochloride, n,n-dimethyl-2-1-piperazinyl nicotinamide hydrochloride, n,n-dimethyl-2-piperazin-1-yl pyridine-3-carboxamide hydrochloride, n,n-dimethyl-2-1-piperazinyl nicotinamide hcl, n,n-dimethyl-2-1-piperazinyl pyridine-3-carboxamide hydrochloride, n,n-dimethyl-2-piperazin-1-yl-pyridine-3-carboxamide hydrochloride, n,n-dimethyl-2-piperazin-1-yl pyridine-3-carboxamide-hydrogen chloride 1/1
PubChem CID 53418086
IUPAC Name N,N-dimethyl-2-piperazin-1-ylpyridine-3-carboxamide;hydrochloride
SMILES CN(C)C(=O)C1=C(N=CC=C1)N2CCNCC2.Cl

Specifications

Quantity 1 g
Formula Weight 270.76
Percent Purity 96%
Chemical Name or Material N,N-Dimethyl-2-(1-piperazinyl)nicotinamide hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.