Learn More
N,N-Diisopropylformamide, 98%
Product Code. 11362756
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362756 25 g 25g
11352756 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11362756 Supplier Thermo Scientific Alfa Aesar Supplier No. L08857.14
 Restricted Item
In Stock
Add to basket
CAS: 2700-30-3 | C7H15NO | 129.203 g/mol
It is employed as a intermediate for organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a intermediate for organic synthesis.
Solubility
Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2700-30-3
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.203
MDL Number MFCD00008867
InChI Key UNBDDZDKBWPHAX-UHFFFAOYSA-N
Synonym n,n-diisopropylformamide, formamide, n,n-bis 1-methylethyl, n-formyldiisopropylamine, diisopropylformamide, n,n-bis 1-methylethyl formamide, nn-di-iso-propylformamide, formamide, n,n-diisopropyl, formamide, bis 1-methylethyl, n,n-bis methylethyl carboxamide, 1y1&nvhy1&1
PubChem CID 75912
IUPAC Name N,N-di(propan-2-yl)formamide
SMILES CC(C)N(C=O)C(C)C
Specifications
Melting Point 10°C to 11°C
Density 0.889
Boiling Point 195°C to 196°C
Flash Point 75°C (167°F)
Refractive Index 1.437
Quantity 25 g
Beilstein 1745406
Solubility Information Slightly soluble in water.
Formula Weight 129.2
Percent Purity 98%
Chemical Name or Material N,N-Diisopropylformamide