Learn More
N,N-diethyl-2-(6-fluoro-1H-indol-3-yl)ethan-1-amine, TRC
Specifications
Specifications
| Chemical Name or Material | 6-fluoro DET |
| CAS | 2836-69-3 |
| Molecular Formula | C14 H19 F N2 |
| Quantity | 100 mg |
| Synonym | 6-FDET, 6-fluoro N,N-DET, 6-fluoro N,N-Diethyltryptamine |
| InChI Formula | InChI=1S/C14H19FN2/c1-3-17(4-2)8-7-11-10-16-14-9-12(15)5-6-13(11)14/h5-6,9-10,16H,3-4,7-8H2,1-2H3 |
| SMILES | CCN(CC)CCc1c[nH]c2cc(F)ccc12 |
| IUPAC Name | N,N-diethyl-2-(6-fluoro-1H-indol-3-yl)ethanamine |
| Molecular Weight (g/mol) | 234.3125 |
| Formula Weight | 234.1532 |