missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dicyclohexylthiourea, 98+%, Thermo Scientific™

Product Code. 11372047
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372047 25 g 25g
11382047 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372047 Supplier Thermo Scientific Alfa Aesar Supplier No. L07785.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1212-29-9
Molecular Formula C13H24N2S
Molecular Weight (g/mol) 240.409
MDL Number MFCD00021316
InChI Key KAJICSGLHKRDLN-UHFFFAOYSA-N
Synonym n,n'-dicyclohexylthiourea, thiourea, n,n'-dicyclohexyl, 1,3-dicyclohexyl-2-thiourea, sym-dicyclohexylthiourea, dicyclohexyl thiourea, sym-dicyclohexythiourea, 1,3-bis cyclohexyl thiourea, n,n'-dicyclohexylthiocarbamide, n,n'-dicyclohexyl-2-thiourea, dicyclohexylthiourea
PubChem CID 727200
IUPAC Name 1,3-dicyclohexylthiourea
SMILES C1CCC(CC1)NC(=S)NC2CCCCC2

Specifications

Melting Point 180°C to 182°C
Odor Odorless
Quantity 25 g
Beilstein 517587
Formula Weight 240.41
Percent Purity ≥98%
Chemical Name or Material N,N'-Dicyclohexylthiourea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.