missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dibenzoylhydrazine, 98+%, Thermo Scientific™

Product Code. 11486077
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486077 5 g 5g
11496077 25 g 25g
11406087 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486077 Supplier Thermo Scientific Alfa Aesar Supplier No. B25081.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 787-84-8
Molecular Formula C14H12N2O2
Molecular Weight (g/mol) 240.262
MDL Number MFCD00003068
InChI Key GRRIYLZJLGTQJX-UHFFFAOYSA-N
Synonym dibenzoylhydrazine, n,n'-dibenzoylhydrazine, 1,2-dibenzoylhydrazine, benzoic acid, 2-benzoylhydrazide, n2-benzoylbenzoic acid hydrazide, hydrazine, 1,2-dibenzoyl, n,n-dibenzoylhydrazine, nn'-dibenzoylhydrazine, c6h5-co-nh-nh-co-c6h5, benzoic acid,2-benzoylhydrazide
PubChem CID 13085
ChEBI CHEBI:38453
IUPAC Name N'-benzoylbenzohydrazide
SMILES C1=CC=C(C=C1)C(=O)NNC(=O)C2=CC=CC=C2

Specifications

Melting Point 238°C to 240°C
Quantity 5 g
Formula Weight 240.26
Percent Purity ≥98%
Chemical Name or Material N,N'-Dibenzoylhydrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.