Learn More
N,N' '-Bis(methylphenyl)-1,4-benzenediamine (Technical Grade), TRC
Specifications
Specifications
| Chemical Name or Material | N,N''-Bis(methylphenyl)-1,4-benzenediamine (Technical Grade) |
| CAS | 27417-40-9 |
| Molecular Formula | C20H20N2 |
| Quantity | 250 mg |
| Synonym | 1,4-Benzenediamine, N,N'-bis(methylphenyl)- (9CI), p-Phenylenediamine, N,N'-ditolyl- (8CI), N1,N4-Bis(methylphenyl)-1,4-benzenediamine, DMPD, N,N’-Bis(methylphenyl)-1,4-benzenediamine, N,N’-Bis(methylphenyl)-p-phenylenediamine, N,N’-Di(monomethylphenyl)-p-phenylenediamine, N,N’-Ditolyl-1,4-phenylenediamine, N,N’-Ditolyl-p-phenylenediamine |
| InChI Formula | InChI=1S/C20H20N2/c1-15-7-3-5-9-19(15)21-17-11-13-18(14-12-17)22-20-10-6-4-8-16(20)2/h3-14,21-22H,1-2H3 |
| SMILES | Cc1ccccc1Nc2ccc(Nc3ccccc3C)cc2 |
| IUPAC Name | 1-N,4-N-bis(2-methylphenyl)benzene-1,4-diamine |
| Molecular Weight (g/mol) | 288.39 |
| Formula Weight | 288.1626 |