Learn More
N,N'-Bis[4-[2-(N-methylcarbamoyl)-4-pyridyloxy]phenyl]urea, TRC
Specifications
Specifications
| Chemical Name or Material | N,N'-Bis[4-[2-(N-methylcarbamoyl)-4-pyridyloxy]phenyl]urea |
| CAS | 284670-98-0 |
| Molecular Formula | C27 H24 N6 O5 |
| Quantity | 100 mg |
| Synonym | 2-Pyridinecarboxamide, 4,4'-[carbonylbis(imino-4,1-phenyleneoxy)]bis[N-methyl-, N-Methyl-4-[4-[[4-[[2-(methylcarbamoyl)-4-pyridyl]oxy]phenyl]carbamoylamino]phenoxy]pyridine-2-carboxamide, N,N'-Bis[4-[2-(N-methylcarbamoyl)-4-pyridyloxy]phenyl]urea, 4,4'-[Carbonylbis(azanediyl-4,1-phenyleneoxy)]bis(Nmethylpyridine-2-carboxamide) Imp. E (EP) |
| InChI Formula | InChI=1S/C27H24N6O5/c1-28-25(34)23-15-21(11-13-30-23)37-19-7-3-17(4-8-19)32-27(36)33-18-5-9-20(10-6-18)38-22-12-14-31-24(16-22)26(35)29-2/h3-16H,1-2H3,(H,28,34)(H,29,35)(H2,32,33,36) |
| SMILES | CNC(=O)c1cc(Oc2ccc(NC(=O)Nc3ccc(Oc4ccnc(c4)C(=O)NC)cc3)cc2)ccn1 |
| IUPAC Name | N-methyl-4-[4-[[4-[2-(methylcarbamoyl)pyridin-4-yl]oxyphenyl]carbamoylamino]phenoxy]pyridine-2-carboxamide |
| Molecular Weight (g/mol) | 512.52 |
| Formula Weight | 512.1808 |