missing translation for 'onlineSavingsMsg'
Learn More

N-Methylperfluorooctanesulfonamide, TRC

Product Code. 30476160
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476160 5 mg 5mg
30472186 25 mg 25mg
30471725 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30476160 Supplier Toronto Research Chemicals Supplier No. TRCM3273405MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31506-32-8
Molecular Formula C9 H4 F17 N O2 S
Molecular Weight (g/mol) 513.17
Synonym 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluoro-N-methyl-1-octanesulfonamide, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluoro-N-methyloctane-1-sulfonamide, AI 3-29758, Fluorad FX 12, N-MeFOSA, N-Methyl perfluorooctanesulphonamide, N-Methylheptadecafluorooctanesulfonamide, N-Methylperfluorooctanesulfonamide, Perfluoro-N-methyloctanesulfonamide, MeFOSA
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N-methyloctane-1-sulfonamide
SMILES CNS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C9H4F17NO2S/c1-27-30(28,29)9(25,26)7(20,21)5(16,17)3(12,13)2(10,11)4(14,15)6(18,19)8(22,23)24/h27H,1H3
Formula Weight 512.9691
Chemical Name or Material N-Methylperfluorooctanesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.