Learn More
N-Methyl-L-serine Monohydrate, TRC
Product Code. 30469341
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469341 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30469341 Supplier Toronto Research Chemicals Supplier No. TRCM30525510MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
CAS 2842007-47-8
Molecular Formula C4H9NO3 . H 2O
Synonym (2S)-3-Hydroxy-2-(methylamino)propanoic Acid Monohydrate
InChI Formula InChI=1S/C4H9NO3.H2O/c1-5-3(2-6)4(7)8;/h3,5-6H,2H2,1H3,(H,7,8);1H2/t3-;/m0./s1
SMILES CN[C@@H](CO)C(=O)O.O
IUPAC Name methyl-L-serine hydrate
Molecular Weight (g/mol) 137.14
Quantity 10 mg
Formula Weight 137.07
Shipping Condition Room Temperature
Chemical Name or Material N-Methyl-L-serine Monohydrate