missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-L-arginine, Thermo Scientific™

Product Code. 15494809
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 15494809

Brand: Thermo Scientific Alfa Aesar J62431.MF

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Methyl-L-arginine is used as a selective inhibitor of L-arginine uptake. It comes under the category of ion pump transporters.

Solubility
Soluble to 100 mM in water

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. For obtaining a higher solubility , please warm the tube at 37°C and shake it in the ultrasonic bath for a while. Stock solution can be stored below -20°C for several months.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2480-28-6
Molecular Formula C7H16N4O2
Molecular Weight (g/mol) 188.231
MDL Number MFCD12828775
InChI Key NTWVQPHTOUKMDI-YFKPBYRVSA-N
Synonym n-methyl-l-arginine, n2-methyl-l-arginine, arginine, n2-methyl-, l, unii-88rbj4ze9r, 88rbj4ze9r, l-monomethyl-l-arginine, s-5-guanidino-2-methylamino pentanoic acid, n 2-methyl-l-arginine, n∼5∼-diaminomethylene-n∼2∼-methylornithine, n-mearg
PubChem CID 73046
IUPAC Name (2S)-5-(diaminomethylideneamino)-2-(methylamino)pentanoic acid
SMILES CNC(CCCN=C(N)N)C(=O)O

Specifications

Color White
Solubility Information Soluble to 100 mM in water
Quantity 50 mg
Formula Weight 188.23
Physical Form Powder
Chemical Name or Material N-Methyl-L-arginine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.