Learn More
N-Methyl-d3-form-d1-amide, CDN
Product Code. 30322691
Change view
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322691 0.25 g 0.25g
30322143 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30322691 Supplier CDN Supplier No. CDND54190.25G
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Methylformamide-D4
CAS 110505-55-0
Molecular Formula C2 D4 H N O
Quantity 0.25 g
Synonym Formamide-1-d, N-(methyl-d3)- (9 CI), N-Methylformamide-D4, N-(Methyl-d3)formamide-1-d, N-Methylformamide-d4
InChI Formula InChI=1 S/C2H5NO/c1-3-2-4/h2H,1H3,(H,3,4)/i1D3,2 D
SMILES [2 H]C(=O)NC([2 H])([2 H])[2 H]
IUPAC Name 1-deuterio-N-(trideuteriomethyl)formamide
Molecular Weight (g/mol) 63.092
Formula Weight 63.062 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity