Learn More
N-Methyl-2-piperidinemethanol-d5, TRC
Product Code. 30483124
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483124 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30483124 Supplier Toronto Research Chemicals Supplier No. TRCM324822250MG
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Methyl-2-piperidinemethanol-d5
CAS 1794971-17-7
Molecular Formula C7H10D5NO
Quantity 250 mg
Synonym (1-Methyl-2-piperidyl)methanol-d5, (1-Methylpiperidin-2-yl)methanol-d5, (±)-2-Hydroxymethyl-1-methylpiperidine-d5, 1-Methyl-2-piperidinemethanol-d5, 2-Hydroxymethyl-1-methylpiperidine-d5, 2-Hydroxymethyl-N-methylpiperidine-d5, NSC 45464-d5
InChI Formula InChI=1S/C7H15NO/c1-8-5-3-2-4-7(8)6-9/h7,9H,2-6H2,1H3/i1D3,6D2
SMILES [2H]C([2H])([2H])N1CCCCC1C([2H])([2H])O
IUPAC Name dideuterio-[1-(trideuteriomethyl)piperidin-2-yl]methanol
Molecular Weight (g/mol) 134.23
Formula Weight 134.1467