missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-2-(1H-1,2,4-triazol-1-ylmethyl)benzylamine, 97%, Thermo Scientific™

Product Code. p-3522599
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11360657 250 mg 250mg
10307454 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11360657 Supplier Thermo Scientific Maybridge Supplier No. CC47546CB
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 906352-64-5
Molecular Formula C11H14N4
Molecular Weight (g/mol) 202.261
MDL Number MFCD09025848
InChI Key VHDVEBIJXKJALO-UHFFFAOYSA-N
Synonym n-methyl-2-1h-1,2,4-triazol-1-ylmethyl benzylamine, methyl 2-1,2,4-triazol-1-ylmethyl phenyl methyl amine, methyl 2-1,2,4-triazolylmethyl phenyl methyl amine, n-methyl-1-2-1h-1,2,4-triazol-1-yl methyl phenyl methanamine
PubChem CID 18525820
IUPAC Name N-methyl-1-[2-(1,2,4-triazol-1-ylmethyl)phenyl]methanamine
SMILES CNCC1=CC=CC=C1CN2C=NC=N2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 202.26
Percent Purity 97%
Chemical Name or Material N-methyl-2-(1H-1, 2, 4-triazol-1-ylmethyl)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.