Learn More
N-Methyl-1H-indole-5-ethanesulfonamide, TRC
Specifications
Specifications
| Chemical Name or Material | N-Methyl-1H-indole-5-ethanesulfonamide |
| CAS | 98623-50-8 |
| Molecular Formula | C11 H14 N2 O2 S |
| Quantity | 25 mg |
| Synonym | N-Methyl-1H-indole-5-ethanesulfonamide, 2-(1H-Indol-5-yl)-N-methylethanesulfonamide |
| InChI Formula | InChI=1S/C11H14N2O2S/c1-12-16(14,15)7-5-9-2-3-11-10(8-9)4-6-13-11/h2-4,6,8,12-13H,5,7H2,1H3 |
| SMILES | CNS(=O)(=O)CCc1ccc2[nH]ccc2c1 |
| IUPAC Name | 2-(1H-indol-5-yl)-N-methylethanesulfonamide |
| Molecular Weight (g/mol) | 238.31 |
| Formula Weight | 238.0776 |